Sodium-2-naphthalenesulphonate ≥99.0% (by HPLC) for ion pair chromatography, Supelco®
Catalog # 70289-10G
Supplier: Merck
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:230.21 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:532-02-5
Formula:C₁₀H₈O₃S
MDL Number:MFCD00064186