

1-(Trimethylsilyl)imidazole : Pyridine (1:4), LiChropur™ for GC derivatisation, Supelco®
: Merck
:8077-35-8
:18156-74-6
:0.98 g/cm³ (20 °C)
:Refrigerator
:1993
:3,II
:M-Clarity™ quality level = MQ100
:17 °C
:GHS02
:GHS07
- Appearance (color):Colorless to Very Faint Yellow
- Density D20/4:0.976 - 0.980
- Refractive Index N20/d:1.502 - 1.504
- Proton NMR Spectrum:Conforms to Structure
Frequently Bought Together