Pesticide standard, Thifensulfuron-methyl, reference material, Honeywell Fluka™
Catalog # HONCR1685-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:79277-27-3
- Description:Thifensulfuron-methyl
- Formula:C₁₂H₁₃N₅O₆S₂
- Storage temperature:Ambient
- Description:Thifensulfuron-methyl
- Form:Powder
Specifications
About this item
High quality organic reference materials produced according to ISO 9001.
Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.