Pesticide standard, Carpropamide, reference material, Honeywell Fluka™
Catalog # HONCR1638-100MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:100 mg
- Standards:Analytical Standard
- CAS No.:104030-54-8
- Description:Carpropamide
- Formula:C₁₅H₁₈Cl₃NO
- Storage temperature:Ambient
- Description:Carpropamide
- Form:Solid
Specifications
About this item
High quality organic reference materials produced according to ISO 9001.
Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.