Pesticide standard, Indoxacarb, reference material, Honeywell Fluka™
HONCR1631-25MG
:
- Pack type:Glass Bottle
- Pk:25 mg
- Standards:Analytical Standard
- CAS No.:144171-61-9
- Description:Indoxacarb
- Formula:C₂₂H₁₇ClF₃N₃O₇
- Storage temperature:Ambient
- Description:Indoxacarb
- Form:Powder
High quality organic reference materials produced according to ISO 9001.
: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.