0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trays/Dishes/Pots
Laboratory Trays/Dishes
Sample pans for Pyris 1 TGA and TGA 8000
Sample pans for Pyris 1 TGA and TGA 8000
Sample pans for Pyris 1 TGA and TGA 8000
PERCN5370464
:
PerkinElmer
Sample pans for Pyris 1 TGA and TGA 8000
PERCN5370464
:
PerkinElmer
For:
TGA 8000 and Pyris 1 TGA
Description:
Ceramic sample pans, pack of 10
Environmentally Preferable:
Pk:
1