Supelco® Organic Reference Standard, EPA 8260 Internal Standards Mix 1
Catalog # 46838-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:2500 µg/ml in methanol
- Pk:1
- Standards:Certified reference material
- Description:EPA 8260 Internal standards mix 1
- Storage conditions:2...30 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370 - H411
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:9 °C
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08, GHS09
Caution: P210 - P273 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311