Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, EPA CLP Semivolatiles Surrogate Standards Mix
Supelco® Organic Reference Standard, EPA CLP Semivolatiles Surrogate Standards Mix
Catalog # 46824-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, EPA CLP Semivolatiles Surrogate Standards Mix
Catalog # 46824-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    4000 µg/ml each component in methylene chloride
  • Pk:
    1 mL
  • Standards:
    Certified reference material
  • Description:
    EPA CLP Semivolatiles surrogate standards mix
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100

Specifications

About this item

  • GHS signal: GHS08, GHS07, GHS09