Order Entry
Switzerland
ContactUsLinkComponent
EPA 8270/Appendix IX nitrosamines mix, certified reference material, Supelco®
EPA 8270/Appendix IX nitrosamines mix, certified reference material, Supelco®
Catalog # 502138.
Supplier:  Merck
CAS Number:  
EPA 8270/Appendix IX nitrosamines mix, certified reference material, Supelco®
Catalog # 502138.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2000 mug/mL each component in methanol
  • Pk:
    1
  • Description:
    EPA 8270/Appendix IX Nitrosamines Mix
  • Type:
    Certified reference material

Specifications

About this item