Supelco® Organic Reference Standard, Phenol-d₆
Catalog # 48716-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:2000 μg/ml in methylene chloride
- Pk:1
- Standards:Certified reference material
- Description:Phenol-d₆
- Storage conditions:2…8 °C
- Hazard class description:H315 - H319 - H336 - H351
- Shipping conditions:STD
- M-Clarity quality level:MQ100
Specifications
About this item
- GHS signal: GHS08, GHS07
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.
Caution: P201 - P202 - P261 - P302 + P352 - P305 + P351 + P338 - P308 + P313