Organic reference standard, Oxypurinol, analytical standard, Supelco®
Catalog # 42688-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:2465-59-0
- Description:Oxypurinol
- Environmentally Preferable:
- Formula:C₅H₄N₄O₂
- Description:Oxypurinol
Specifications
About this item
- Biochemical/physiol action: An allopurinol metabolite