Order Entry
Switzerland
ContactUsLinkComponent
Pesticide reference standard, N-Desmethylthiamethoxam, analytical standard, Supelco®
Pesticide reference standard, N-Desmethylthiamethoxam, analytical standard, Supelco®
Catalog # 73348-50MG
Supplier:  Merck
CAS Number:  
undefined
Pesticide reference standard, N-Desmethylthiamethoxam, analytical standard, Supelco®
Catalog # 73348-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    171103-04-1
  • Description:
    N-Desmethylthiamethoxam
  • Formula:
    C₇H₈ClN₅O₃S
  • Description:
    N-Desmethylthiamethoxam

Specifications

About this item

Packaging: Bottomless glass bottle contents are inside inserted fused cone.