Organic reference standard, Isoamyl 4-methoxycinnamate, analytical standard, Supelco®
Katalognr. 44189-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:71617-10-2
- Description:Isoamyl 4-methoxycinnamate
- Formula:C₁₅H₂₀O₃
- Description:Isoamyl 4-methoxycinnamate
Specifications
About this item
Isoamyl 4-methoxycinnamate may be used as a reference standard in the determination of isoamyl 4-methoxycinnamate in water samples using liquid chromatography coupled with ultraviolet spectrophotometry detector (LC-UV).
- Derivative of cinnamon acid ester
- Utilised as a sunscreen agent
- Act as a waterproof filter for ultraviolet-B (UV-B) rays