Order Entry
Switzerland
ContactUsLinkComponent
 
Organic reference standard, Iscotrizinol, analytical standard, Supelco®
  05846-25MG
 :  Merck
 :  
undefined
Organic reference standard, Iscotrizinol, analytical standard, Supelco®
  05846-25MG
 :  Merck
 :  

 

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    154702-15-5
  • Description:
    Iscotrizinol
  • Formula:
    C₄₄H₅₉N₇O₅
  • Description:
    Iscotrizinol

 

 

 : Bottomless glass bottles are inserted with fused cone.