Order Entry
Switzerland
ContactUsLinkComponent
Pesticide reference standard, Dimetilan, PESTANAL®, analytical standard, Supelco®
Pesticide reference standard, Dimetilan, PESTANAL®, analytical standard, Supelco®
Catalog # 45450-50MG
Supplier:  Merck
CAS Number:  
undefined
Pesticide reference standard, Dimetilan, PESTANAL®, analytical standard, Supelco®
Catalog # 45450-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Analytical standard
  • CAS No.:
    644-64-4
  • Description:
    Dimetilan
  • Formula:
    C₁₀H₁₆N₄O₃
  • Description:
    Dimetilan

Specifications

About this item