Organic reference standard, Benzophenone-9, analytical standard, Supelco®
Benzophenone-9, Analytical standard, 25 mg
Catalog # 94257-25MG
Supplier: Merck
Organic reference standard, Benzophenone-9, analytical standard, Supelco®
Catalog # 94257-25MG
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:25 mg
- Standards:Analytical standard
- Description:Benzophenone-9
- Cat. No.:94257-25MG
- Formula:C₁₅H₁₂Na₂O₁₁S₂
- Description:Benzophenone-9
- CAS No.:76656-36-5
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



