Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standard, 3-Acetamidophenol, analytical standard, Supelco®
Organic reference standard, 3-Acetamidophenol, analytical standard, Supelco®
Catalog # 19374-100MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 3-Acetamidophenol, analytical standard, Supelco®
Catalog # 19374-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    621-42-1
  • Description:
    3-Acetamidophenol
  • Melting point:
    145...148 °C (lit.)
  • Formula:
    C₈H₉NO₂
  • Description:
    3-Acetamidophenol

Specifications

About this item