Organic reference standard, 1-Phenyloctadecane, analytical standard, Supelco®
Catalog # 74753-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Analytical standard
- CAS No.:4445-07-2
- Description:1-Phenyloctadecane ≥99,5%
- Melting point:35...37 °C
- Formula:C₆H₅(CH₂)₁₇CH₃
- Description:1-Phenyloctadecane
- Boiling point:408 °C (lit)
Specifications
About this item
Packaging: Packed in bottomless glass bottle. Contents are inside inserted fused cone.