Order Entry
Switzerland
ContactUsLinkComponent
Supelco® Organic Reference Standard, (±)-α-Hexabromocyclododecane
Supelco® Organic Reference Standard, (±)-α-Hexabromocyclododecane
Catalog # 30373-25MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, (±)-α-Hexabromocyclododecane
Catalog # 30373-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    134237-50-6
  • Description:
    (±)-α-Hexabromocyclododecane
  • Environmentally Preferable:
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H361 - H362 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD28118917
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    α-HBCD; (1RS,2RS,5SR,6RS,9RS,10SR)-1,2,5,6,9,10-Hexabromocyclododecane
  • Formula:
    C₁₂H₁₈Br₆
  • Molecular weight:
    641,70
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS08, GHS09
Caution: P202 - P260 - P263 - P264 - P273 - P308 + P313