Order Entry
Switzerland
ContactUsLinkComponent
 
Restriction enzymes, FastDigest SdaI, Fermentas
  FD1194
 :  
undefined
Restriction enzymes, FastDigest SdaI, Fermentas
  FD1194
 :  

 

  • Pk:
    1 KIT
  • Compatible buffer:
    FastDigest Green Buffer or FastDigest Buffer (colorless)
  • Overhang type:
    3' overhang
  • Description:
    FastDigest SdaI
  • Enzyme name:
    FastDigest SdaI
  • No. of reactions:
    20 (20 µl)

 

 

FastDigest SdaI is an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest green reaction buffers. This enzyme recognises CCTGCA^GG site and cuts best at 37 °C in 5 to 15 minutes using universal FastDigest buffer.

  • Direct loading on gels and 176 FastDigest enzymes available
  • No star activity
  • Buffer compatibility of 100% with downstream applications
  • Methylation sensitivity: not CpG methylation-sensitive, dam methylation-sensitive and dcm methylation-sensitive

The universal buffer allows rapid single-, double-, or multiple DNA digestion within 5 to 15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps.

Applications include molecular cloning, restriction site mapping, genotyping, southern blot, restriction fragment length polymorphism (RFLP) and SNP analysis.

 : For research use only. Not for use in diagnostic procedures.