ContactUsLinkComponent
Gas burette, Bunte, Class A
Gas burette, Bunte, Class A
  WITG2.970.000
undefined
Gas burette, Bunte, Class A
  WITG2.970.000
  • Capacity:
    100 ml
  • Division:
    0,2 ml
  • Pk:
    1
Gas burette made of borosilicate glass, accuracy Class A.
  • With resistant DIFFICO graduation
  • Main point ring graduation
ST-stopcock with double bore and screw-thread retaining nut.