Silver sulphate ≥99.0% (by argentometric titration) ACS for analysis
Supplier: PanReac AppliChem
Certificates
About this item
CAS Number:10294-26-5
:
Formula:Ag₂SO₄
MDL Number:MFCD00003407
:311.8 g/mol
Melting Pt:652 °C
Storage Temperature:Ambient
EINECS:233-653-7
Specification Test Results
- Minimum Assay (Argentometric):99.0%
- Identity:Passes Test
- Suitability: For COD Determination According to UNE 77-004-89:Passes Test
- Insoluble Matter in H₂O:≤0.02%
- Chloride (Cl):≤0.001%
- Non-Precipitated Substances by HCl (as SO₄):≤0.03%
- Nitrate (NO₃):≤0.001%
- Ca:≤0.002%
- Cd:≤0.0005%
- Co:≤0.0005%
- Cu:≤0.0005%
- Fe:≤0.001%
- K:≤0.005%
- MG:≤0.002%
- NA:≤0.005%
- Ni:≤0.0005%
- PB:≤0.0005%
- Zn:≤0.0005%
Specification Test Results
Frequently Bought Together