Colorimetric rapid tests, sulphate, VISOCOLOR® ECO
Supplier: MACHEREY-NAGEL
Certificates
About this item
Reagents for the determination of sulfate in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
Remarks: Measuring range on PF-12Plus. Range on other photometers can be different
Suitable for sea water testing after dilution.
Specifications
- Platform:Turbidity-based test kits
- Storage temperature:15...25 °C
- Shelf life (months):36
- Weight:254 g (Including packaging)