ContactUsLinkComponent
Dispensing pins, MICRO PIN®
Dispensing pins, MICRO PIN®
Catalog # BRAU415019
Supplier:  B. Braun
Dispensing pins, MICRO PIN®
Catalog # BRAU415019
Supplier:  B. Braun
Specifications
  • Description:
    MICRO PIN® non–vented, sterile dispensing pin
  • Pk:
    100
Specifications
About this item
Single use dispensing pin is used for withdrawal or injection of medication from rubber-stopper vials and bag injection ports.
  • Compatible with Luer lock connection
  • Simple design with large internal lumen
  • Eliminates the risk of needle stick injuries
  • High fluid-flow rate and comfortable easy handling
Made of plastic and free from PVC, DEHP and natural rubber latex.