ContactUsLinkComponent
Wright's stain solution
 :  Merck
Wright's stain solution
 :  Merck
  • Suitability for microscopy (Blood smear):
    passes test
  • Erythrocytes:
    pink to brownish
  • Nuclei:
    blue-violet
  • Eosinophilic granules:
    red to red-brown
  • Neutrophilic granules:
    light violet
  • Lymphocyte cytoplasm:
    Blue
Frequently Bought Together