Supelco® Organic Reference Standard, Dextran
Catalog # 31430-1EA
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Standards:Analytical standard, for GPC, Set Mp 1000-400000
- CAS No.:9004-54-0
- Description:Dextran
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00130935
- M-Clarity quality level:MQ100
- Formula:[C₆H₁₀O₅]ₙ
- Pk Info:10 standards
Specifications
About this item
Certifications: Exact values can be taken from the accompanying certificate of analysis.