Order Entry
Switzerland
ContactUsLinkComponent
Test strips, qualitative, fluoride
Test strips, qualitative, fluoride
Catalog # MANA90750
Supplier:  MACHEREY-NAGEL
CAS Number:  
undefined
Test strips, qualitative, fluoride
Catalog # MANA90750
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Alizarin red
  • Pk:
    200
  • Description:
    Fluoride test papers

Specifications

About this item

The test paper allows the rapid detection of fluoride ions in solutions containing hydrochlorid acid.

  • Colour change: Pink to yellow-white

It is suitable for the safe and easy detection of dangerous hydrofluoric acid, which is used for example in computer chip production.

Limit of sensitivity: 20 mg/L F⁻

Packaging: Pack of 200 strips 20×70 mm