Order Entry
Switzerland
ContactUsLinkComponent
Lithium dodecyl sulphate ≥98.5%, HiPerSolv CHROMANORM® for HPLC
Flammable
Corrosive
Irritant
Lithium dodecyl sulphate ≥98.5%, HiPerSolv CHROMANORM® for HPLC
Catalog # 85793.150
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Corrosive
Irritant
Lithium dodecyl sulphate ≥98.5%, HiPerSolv CHROMANORM® for HPLC
Catalog # 85793.150
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Assay (C12):
    ≥ 98,50 %
  • Appearance:
    White fine crystals
  • IR Spectrum:
    Passes test
  • Loss on drying (120°C; under vacuum):
    ≤ 0,50 %
  • Appearance of solution (1 %; ethanol):
    Clear and colourless
  • Appearance of solution (3 %):
    Clear and colourless
  • Cl (Chloride):
    ≤ 100,0 ppm
  • Absorbance (260 nm) (3 %):
    ≤ 0,100
  • Absorbance (280 nm) (3 %):
    ≤ 0,075

Specification Test Results

About this item

 :
 :272.34 g/mol
Storage Temperature:Ambient
UN:1325
ADR:4.1,III
 :GHS02
 :GHS05
 :GHS07
CAS Number:2044-56-6
Formula:C₁₂H₂₅LiO₄S
MDL Number:MFCD00007467