Haematoxylin hydrate, Gurr™ for microscopical staining
Catalog # 340374T
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Dye content (spectrophotometrically):Min 95 %
- Identity (UV/VIS-Spectrum):passes test
- Absorption maximum l max (acetonitrile):290 - 292 nm
- Spec. Absorptivity A 1%/1cm:155 - 165
- Haematein (spectrophotometrically):Max 0.1 %
Specification Test Results
About this item
:
:302.29 g/mol (anhy)
:1
CAS Number:517-28-2
:208-237-3
Formula:C₁₆H₁₄O₆·xH₂O
MDL Number:MFCD00078111