Order Entry
Switzerland
ContactUsLinkComponent
Organic analytical standards, Rhodamine B-d20, neat, reference material, Honeywell Fluka™
Organic analytical standards, Rhodamine B-d20, neat, reference material, Honeywell Fluka™
Catalog # HONCR1513-10MG
CAS Number:  
Organic analytical standards, Rhodamine B-d20, neat, reference material, Honeywell Fluka™
Catalog # HONCR1513-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • CAS No.:
    1610999-57-9
  • Description:
    Rhodamine B-d₂0
  • Formula:
    C₂₈H₁₁D₂₀ClN₂O₃
  • Storage temperature:
    Ambient
  • Description:
    Rhodamine B-d₂0
  • Freezing point:
    > 190 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results