Organic analytical standards, Brilliant green-d5 hydrogen sulfate hydrate, neat, reference material, Honeywell Fluka™
Catalog # HONCR1496-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Packung:Glass bottle
- VE:10 mg
- Bezeichnung:Brilliant green-d₅ hydrogen sulfate hydrate
- Formel:C₂₇H₂₈D₅N₂ · HSO₄
- Lagertemperatur:Ambient
- Bezeichnung:Brilliant green-d₅ hydrogen sulfate hydrate
- Gefrierpunkt:215...220 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results