Pesticide reference standard, 2’,4’-Formoxylidide-d6, neat, Honeywell Fluka™
Catalog # HONCR1478-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- Description:2’,4’-Formoxylidide-d₆
- Formula:C₉H₅D₆NO
- Storage temperature:Ambient
- Description:2’,4’-Formoxylidide-d₆
- Freezing point:115...117 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results