Pharmaceutical analytical standard, m-Chloramphenicol erythro form, reference material, Honeywell Fluka™
Catalog # HONCR1471-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:138125-71-0
- Description:m-Chloramphenicol erythro form
- Formula:C₁₁H₁₂Cl₂N₂O₅
- Storage temperature:Ambient
- Description:m-Chloramphenicol erythro form
- Freezing point:134...136 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results