Order Entry
Switzerland
ContactUsLinkComponent
Pharmaceutical analytical standard, Lomefloxacin-d5 hydrochloride, reference material, Honeywell Fluka™
Pharmaceutical analytical standard, Lomefloxacin-d5 hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1430-10MG
CAS Number:  
Pharmaceutical analytical standard, Lomefloxacin-d5 hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1430-10MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mg
  • Description:
    Lomefloxacin-d₅ hydrochloride
  • Formula:
    C₁₇H₁₄D₅F₂N₃O₃ · HCl
  • Storage temperature:
    Ambient
  • Description:
    Lomefloxacin-d₅ hydrochloride
  • Freezing point:
    >310 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results