Pharmaceutical analytical standard, Mapenterol hydrochloride, reference material, Honeywell Fluka™
Catalog # HONCR1402-10MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:54238-51-6
- Description:Mapenterol hydrochloride
- Formula:C₁₄H₂₀ClF₃N₂O · HCl
- Storage temperature:Ambient
- Description:Mapenterol hydrochloride
- Freezing point:176...181 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results