Pharmaceutical analytical standard, Menidazole, reference material, Honeywell Fluka™
Katalognr. HONCR1345-10MG
Anbieter: Honeywell Chemicals
CAS-Nummer:
Technische Daten
- Pack type:Glass bottle
- Pk:10 mg
- CAS No.:696-23-1
- Description:Menidazole
- Formula:C₄H₅N₃O₂
- Storage temperature:Ambient
- Description:Menidazole
- Flash point:195.4 °C
- Freezing point:252...257 °C
Technische Daten
Über diesen Artikel
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results