Order Entry
Switzerland
ContactUsLinkComponent
Pesticide reference standard, Fipronil sulfone, neat, Honeywell Fluka™
Pesticide reference standard, Fipronil sulfone, neat, Honeywell Fluka™
  HONCR1327-50MG
 :  
Pesticide reference standard, Fipronil sulfone, neat, Honeywell Fluka™
  HONCR1327-50MG
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    50 mg
  • CAS No.:
    120068-36-2
  • Description:
    Fipronil sulfone
  • Formula:
    C₁₂H₄Cl₂F₆N₄O₂S
  • Storage temperature:
    Ambient
  • Description:
    Fipronil sulfone
  • Freezing point:
    200 °C

 

 

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results