Pesticide reference standard, Crufomate, neat, Honeywell Fluka™
Catalog # HONCR1169-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 mg
- CAS No.:299-86-5
- Description:Crufomate
- Formula:C₁₂H₁₉ClNO₃P
- Storage temperature:Ambient
- Description:Crufomate
- Freezing point:55...59 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results