Pesticide reference standard, Phenmedipham-ethyl, neat, Honeywell Fluka™
Catalog # HONCR1133-25MG
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Packung:Glass bottle
- VE:25 mg
- CAS Nr.:13684-44-1
- Bezeichnung:Phenmedipham-ethyl
- Formel:C₁₇H₁₈N₂O₄
- Lagertemperatur:Ambient
- Bezeichnung:Phenmedipham-ethyl
- Gefrierpunkt:140...145 °C
Specifications
About this item
High quality inorganic and organic reference materials produced according to ISO 9001.
- Ensure you have the information you need to quickly start your analysis, and pass relevant audits
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results