Order Entry
Switzerland
ContactUsLinkComponent
Pesticide reference standard, Pyridalyl, neat, Honeywell Fluka™
Pesticide reference standard, Pyridalyl, neat, Honeywell Fluka™
Catalog # HONCR1124-25MG
CAS Number:  
Pesticide reference standard, Pyridalyl, neat, Honeywell Fluka™
Catalog # HONCR1124-25MG
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mg
  • CAS No.:
    179101-81-6
  • Description:
    Pyridalyl
  • Formula:
    C₁₈H₁₄Cl₄F₃NO₃
  • Storage temperature:
    Ambient
  • Description:
    Pyridalyl
  • Boiling point:
    227 °C

Specifications

About this item

High quality inorganic and organic reference materials produced according to ISO 9001.

  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results