ContactUsLinkComponent
Wax plates for sealing capillary tubes
Wax plates for sealing capillary tubes
Catalog # MARI2960409
Supplier:  MARIENFELD
undefined
Wax plates for sealing capillary tubes
Catalog # MARI2960409
Supplier:  MARIENFELD
Specifications
  • Description:
    Wax plates for sealing capillary tubes
  • Environmentally Preferable:
  • Pk:
    6
Specifications
About this item
Wax plates are used for sealing capillary tubes for micro hematocrits.
  • Two deposit areas on a plastic base
  • Numbered up to 48
Contain modelling clay and additives of food filler material and natural bees wax. We consciously forego adding any chemicals which could affect the plastic material after some time.
Packaging: Packed in poly bags.