VISOCOLOR® Colorimetric Test, Nitrate
Reagents for the determination of nitrate in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
- Model:MANA931041
- Platform:Colorimetric test kits
- Storage temperature:15…25 °C
- Shelf life (months):18
- Weight:202,74 g (including packaging)
Frequently Bought Together