VISOCOLOR® Colorimetric Test, Acidity
Catalog # MANA915006
Supplier: MACHEREY-NAGEL
CAS Number:
Specifications
- Chemical method:Phenolphthalein
- Pk:1 KIT
- Description:VISOCOLOR® HE Acidity AC 7
- Dimensions:148×122×40 mm (Including packaging)
- Platform:Titrimetric test kits
- Storage temperature:15...25 °C
- Shelf life (months):24
- Weight:233,4 g (Including packaging)
- For unit:Visual
- Tests per kit:200
- Range:0,2 - 7,2 - mmol/L (H⁺) - (1 syringe filling); 0,2 - 7,2 - mmol/L (C(O₂)) - (1 syringe filling)
Specifications
About this item
Reagents for the determination of acidity in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
Delivery:
Reagents for 200 tests, titration syringe and accessories in a box.