ContactUsLinkComponent
Colorimetric rapid tests, phosphate, VISOCOLOR®
Colorimetric rapid tests, phosphate, VISOCOLOR®
Nome del fornitore:  MACHEREY-NAGEL
Certificati
undefined
Colorimetric rapid tests, phosphate, VISOCOLOR®
Nome del fornitore:  MACHEREY-NAGEL
Specifiche
  • Platform:
    Colorimetric test kits
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    24
  • Weight:
    99,6 g (Including packaging)
Specifiche
Frequently Bought Together