VISOCOLOR® Colorimetric Test, Sulphite
Supplier: MACHEREY-NAGEL
Certificates
About this item
Reagents for the determination of sulphite in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
Specifications
- Model:MANA915208
- Platform:Titrimetric test kits
- Storage temperature:15...25 °C
- Shelf life (months):36
- Weight:388,5 g (Including packaging)