ContactUsLinkComponent
VISOCOLOR® Colorimetric Test, Ammonia
VISOCOLOR® Colorimetric Test, Ammonia
Supplier:  MACHEREY-NAGEL
Certificates
undefined
VISOCOLOR® Colorimetric Test, Ammonia
Supplier:  MACHEREY-NAGEL
Certificates
Specifications
  • Model:
    MANA935012
  • Platform:
    Colorimetric test kits
  • Storage temperature:
    15...25 °C
  • Shelf life (months):
    18
  • Weight:
    110,5 g (Including packaging)