VISOCOLOR® Colorimetric Tests, Chlorine
Reagents for the determination of chlorine in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
- Model:MANA931219
- Platform:Photometric test kits
- Storage temperature:15...25 °C
- Shelf life (months):24
- Weight:145,1 g (Including packaging)
IFU Colorimetric Test Kit VISOCOLOR Alpha Chlorine.pdf IFU Colorimetric Test Kit VISOCOLOR ECO Chlorine 1 Free and Total.pdf IFU Colorimetric Test Kit VISOCOLOR ECO Chlorine 2.pdf IFU Colorimetric Test Kit VISOCOLOR ECO Chlorine 6.pdf IFU Colorimetric Test Kit VISOCOLOR ECO Swimming pool Chlorine pH.pdf
More
Frequently Bought Together