ContactUsLinkComponent
Cell-based assay calcein AM
Cell-based assay calcein AM
Catalog # CAYM400146-1
Supplier:  Cayman Chemical
CAS Number:  
Cell-based assay calcein AM
Catalog # CAYM400146-1
Supplier:  Cayman Chemical
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Cell-based assay calcein AM
Specifications
About this item
Packaging: This vial contains 50 µl of 1 mM calcein AM solution in DMSO.