AZOCOLL™ substrate, Calbiochem®
Catalog # 194932-1
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pk:1 g
- Description:AZOCOLL™ substrate
Specifications
About this item
General proteolytic substrate. Provides a rapid, simple method for the detection of proteases in aqueous solutions.
- Colour yield (520 nm): >1,0
- Blank (520 nm, 50 mg/5 ml buffer, 10 minutes at 37 °C, pH 7,0): <0,2
- Mesh: <50 (minimum 80%)
Packaging: Packed in plastic ampoule.