Order Entry
Switzerland
ContactUsLinkComponent
Sodium tetrakis(4-fluorophenyl)borate dihydrate 98% (by HPLC)
Irritant
Sodium tetrakis(4-fluorophenyl)borate dihydrate 98% (by HPLC)
Certificates
Irritant
Sodium tetrakis(4-fluorophenyl)borate dihydrate 98% (by HPLC)
Certificates

Specification Test Results

  • Assay (HPLC):
    Min. 97.5%

Specification Test Results

Related Information

Frequently Bought Together